6-Iodoimidazo[1,2-a]pyridine-2-carboxylic acid 95%

Lieferant: Apollo Scientific

OR32662-1G OR32662-5G
APOSOR32662-1GEA 244 EUR
APOSOR32662-1G APOSOR32662-5G
6-Iodoimidazo[1,2-a]pyridine-2-carboxylic acid 95%
6-Iodoimidazo[1,2-a]pyridine-2-carboxylic acid
Formel: C₈H₅IN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02186210
CAS-Nummer: 478040-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR