3-(Dimethylamino)-1-(2-pyrazinyl)-2-propen-1-one 95%
Lieferant: Apollo Scientific
Synonyme:
3-(Dimethylamino)-1-(pyrazin-2-yl)prop-2-en-1-one
OR32689-1G
OR32689-5G
APOSOR32689-1GEA
538
EUR
APOSOR32689-1G
APOSOR32689-5G
3-(Dimethylamino)-1-(2-pyrazinyl)-2-propen-1-one 95%
3-(Dimethylamino)-1-(2-pyrazinyl)-2-propen-1-one
Formel:
C₉H₁₁N₃O Molecular Weight: 177,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02325230 CAS-Nummer: 111781-53-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...