2-Brom-5-hydroxybenzaldehyd

Lieferant: Apollo Scientific
Achtung

OR3270-100G OR3270-25G OR3270-5G
APOSOR3270-100GEA 95.7 EUR
APOSOR3270-100G APOSOR3270-25G APOSOR3270-5G
2-Brom-5-hydroxybenzaldehyd
2-Bromo-5-hydroxybenzaldehyde
Formel: C₇H₅BrO₂
Molecular Weight: 201.02 g/mol
Schmelzpunkt: 130…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03033065
CAS-Nummer: 2973-80-0
UN: 3077
ADR: 9+EHS III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR