1-(3-Aminophenyl)pyrrolidin-2-one 95%
Lieferant: Apollo Scientific
OR32702-1G
APOSOR32702-1GEA
180
EUR
APOSOR32702-1G
1-(3-Aminophenyl)pyrrolidin-2-one 95%
1-(3-Aminophenyl)pyrrolidin-2-one
Formel:
C₁₀H₁₂N₂O Schmelzpunkt: 29…32 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00468694 CAS-Nummer: 31992-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...