4-Aminochinazolin 95%

Lieferant: Apollo Scientific

Synonyme: Quinazolin-4-amine, Quinazolin-4-ylamine

OR32708-1G OR32708-5G
APOSOR32708-1GEA 62.5 EUR
APOSOR32708-1G APOSOR32708-5G
4-Aminochinazolin 95%
4-Aminoquinazoline
Formel: C₈H₇N₃
Molecular Weight: 145,06 g/mol
Schmelzpunkt: 267…274 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00092016
CAS-Nummer: 15018-66-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR