2-Brom-5-formylpyridin 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromonicotinaldehyde, 2-Bromopyridine-5-carbaldehyde, 2-Bromopyridine-5-carboxaldehyde, 6-Bromo-3-pyridinecarboxaldehyde
OR3271-100G
OR3271-25G
OR3271-5G
APOSOR3271-100GEA
171
EUR
APOSOR3271-100G
APOSOR3271-25G
APOSOR3271-5G
2-Brom-5-formylpyridin 99%
2-Brom-5-formylpyridin
Formel:
C₆H₄BrNO Molecular Weight: 186,01 g/mol Schmelzpunkt: 104…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115419 CAS-Nummer: 149806-06-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...