4'-Chloracetophenonoxim 95%

Lieferant: Apollo Scientific

Synonyme: 1-(4-Chlorophenyl)-1-ethanone oxime

OR32714-1G OR32714-25G OR32714-5G
APOSOR32714-1GEA 180 EUR
APOSOR32714-1G APOSOR32714-25G APOSOR32714-5G
4'-Chloracetophenonoxim 95%
4'-Chloracetophenonoxim
Formel: C₈H₈ClNO
Molecular Weight: 169,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02609517
CAS-Nummer: 1956-39-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR