6-Chloro-2H-chromene-3-carbonitrile 95%
Lieferant: Apollo Scientific
OR32728-1G
OR32728-5G
APOSOR32728-1GEA
270
EUR
APOSOR32728-1G
APOSOR32728-5G
6-Chloro-2H-chromene-3-carbonitrile 95%
6-Chloro-2H-chromene-3-carbonitrile
Formel:
C₁₀H₆ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052359 CAS-Nummer: 57543-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...