N-(4-Chlorophenyl)-2-methyl-2-oxiranecarboxamide 95%
Lieferant: Apollo Scientific
OR32747-1G
OR32747-5G
APOSOR32747-1GEA
336
EUR
APOSOR32747-1G
APOSOR32747-5G
N-(4-Chlorophenyl)-2-methyl-2-oxiranecarboxamide 95%
N-(4-Chlorophenyl)-2-methyl-2-oxiranecarboxamide
Formel:
C₁₀H₁₀ClNO₂ Molecular Weight: 211,65 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00232082 CAS-Nummer: 65925-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...