4-[(Dimethylamino)methylene]-2-phenyl-1,3-oxazol-5(4H)-one 95%

Lieferant: Apollo Scientific

Synonyme: (4E)-4-[(Dimethylamino)methylidene]-2-phenyl-4,5-dihydro-1,3-oxazol-5-one

OR32757-1G OR32757-5G
APOSOR32757-1GEA 270 EUR
APOSOR32757-1G APOSOR32757-5G
4-[(Dimethylamino)methylene]-2-phenyl-1,3-oxazol-5(4H)-one 95%
4-[(Dimethylamino)methylene]-2-phenyl-1,3-oxazol-5(4H)-one
Formel: C₁₂H₁₂N₂O₂
Molecular Weight: 216,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00832526
CAS-Nummer: 51254-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR