3-(tert-Butyl)-3,4-dihydro-2H-1,4-benzoxazine 95%

Lieferant: Apollo Scientific

OR32764-1G OR32764-5G
APOSOR32764-1GEA 336 EUR
APOSOR32764-1G APOSOR32764-5G
3-(tert-Butyl)-3,4-dihydro-2H-1,4-benzoxazine 95%
3-(tert-Butyl)-3,4-dihydro-2H-1,4-benzoxazine
Formel: C₁₂H₁₇NO
Molecular Weight: 191.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01936013
CAS-Nummer: 32278-16-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR