Ethyl-1-(5-formyl-2-pyridinyl)-4-piperidinecarboxylate 95%

Lieferant: Apollo Scientific

OR32796-1G OR32796-5G
APOSOR32796-1GEA 306 EUR
APOSOR32796-1G APOSOR32796-5G
Ethyl-1-(5-formyl-2-pyridinyl)-4-piperidinecarboxylate 95%
Ethyl-1-(5-formyl-2-pyridinyl)-4-piperidinecarboxylate
Formel: C₁₄H₁₈N₂O₃
Molecular Weight: 262,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04973727
CAS-Nummer: 886360-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR