4-[(Dimethylamino)methylene]-2,4-dihydro-3H-pyrazol-3-one 95%
Lieferant: Apollo Scientific
OR32815-1G
OR32815-250MG
APOSOR32815-1GEA
329
EUR
APOSOR32815-1G
APOSOR32815-250MG
4-[(Dimethylamino)methylene]-2,4-dihydro-3H-pyrazol-3-one 95%
4-[(Dimethylamino)methylene]-2,4-dihydro-3H-pyrazol-3-one
Formel:
C₆H₉N₃O Molecular Weight: 139,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00138484 CAS-Nummer: 821016-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...