1-[(Ammoniooxy)methyl]-2,4-dichlorbenzolchlorid 95%
Lieferant: Apollo Scientific
Synonyme:
O-(2,4-Dichlorobenzyl)hydroxylamine hydrochloride, O-[(2,4-Dichlorophenyl)methyl]hydroxylamine hydrochloride
OR32823-1G
OR32823-5G
APOSOR32823-1GEA
180
EUR
APOSOR32823-1G
APOSOR32823-5G
1-[(Ammoniooxy)methyl]-2,4-dichlorbenzolchlorid 95%
1-[(Ammoniooxy)methyl]-2,4-dichlorbenzolchlorid
Formel:
C₇H₈Cl₃NO Molecular Weight: 228,5000 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01114586 CAS-Nummer: 51572-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...