4-[(Ammoniooxy)methyl]-1,2-dichlorbenzolchlorid 95%

Lieferant: Apollo Scientific

Synonyme: O-[(3,4-Dichlorophenyl)methyl]hydroxylamine hydrochloride, O-(3,4-Dichlorobenzyl)hydroxylamine hydrochloride

OR32824-1G OR32824-5G
APOSOR32824-1GEA 336 EUR
APOSOR32824-1G APOSOR32824-5G
4-[(Ammoniooxy)methyl]-1,2-dichlorbenzolchlorid 95%
4-[(Ammoniooxy)methyl]-1,2-dichlorbenzolchlorid
Formel: C₇H₈Cl₃NO
Molecular Weight: 228.5035 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01114587
CAS-Nummer: 15256-10-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR