1-(3,4-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: 1-[(3,4-Dichlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid

OR32827-1G OR32827-5G OR32827-25G
APOSOR32827-1GEA 122 EUR
APOSOR32827-1G APOSOR32827-5G APOSOR32827-25G
1-(3,4-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%
1-(3,4-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Formel: C₁₃H₉Cl₂NO₃
Molecular Weight: 298.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00139964
CAS-Nummer: 64488-03-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR