5-Chloro-4-methyl-1,2,3-thiadiazole 95%
Lieferant: Apollo Scientific
OR32828-1G
OR32828-5G
APOSOR32828-1GEA
336
EUR
APOSOR32828-1G
APOSOR32828-5G
5-Chloro-4-methyl-1,2,3-thiadiazole 95%
5-Chloro-4-methyl-1,2,3-thiadiazole
Formel:
C₃H₃ClN₂S Molecular Weight: 134.59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00202055 CAS-Nummer: 53645-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...