1-Benzyl-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%

Lieferant: Apollo Scientific

OR32832-1G OR32832-5G OR32832-25G
APOSOR32832-1GEA 180 EUR
APOSOR32832-1G APOSOR32832-5G APOSOR32832-25G
1-Benzyl-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%
1-Benzyl-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Formel: C₁₃H₁₁NO₃
Molecular Weight: 229.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00139999
CAS-Nummer: 89960-36-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR