1-(3,4-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: 1-[(3,4-Dichlorophenyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid

OR32835-1G OR32835-5G
APOSOR32835-1GEA 163 EUR
APOSOR32835-1G APOSOR32835-5G
1-(3,4-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%
1-(3,4-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid
Formel: C₁₃H₉Cl₂NO₃
Molecular Weight: 298,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00139865
CAS-Nummer: 338755-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR