6-Methoxy-2H-chromen-3-carbonsäure 95%

Lieferant: Apollo Scientific

OR32844-1G OR32844-25G OR32844-5G
APOSOR32844-1GEA 122 EUR
APOSOR32844-1G APOSOR32844-25G APOSOR32844-5G
6-Methoxy-2H-chromen-3-carbonsäure 95%
6-Methoxy-2H-chromen-3-carbonsäure
Formel: C₁₁H₁₀O₄
Molecular Weight: 206,1947 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00202057
CAS-Nummer: 57543-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR