2-(3-Chlorophenyl)-3-(dimethylamino)acrylaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: (2Z)-2-(3-Chlorophenyl)-3-(dimethylamino)prop-2-enal

OR32855-1G OR32855-5G
APOSOR32855-1GEA 270 EUR
APOSOR32855-1G APOSOR32855-5G
2-(3-Chlorophenyl)-3-(dimethylamino)acrylaldehyde 95%
2-(3-Chlorophenyl)-3-(dimethylamino)acrylaldehyde
Formel: C₁₁H₁₂ClNO
Molecular Weight: 209.68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125587
CAS-Nummer: 86723-63-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR