Nicotinaldehydoxim 95%
Lieferant: Apollo Scientific
Synonyme:
3-Pyridinealdoxime, Pyridine-3-carboxaldoxime, Pyridine-3-aldoxime
OR32892-25G
APOSOR32892-25GEA
126
EUR
APOSOR32892-25G
Nicotinaldehydoxim 95%
Nicotinaldehydoxim
Formel:
C₆H₆N₂O Molecular Weight: 122,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006409 CAS-Nummer: 1193-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...