2-Methyl-2-propanyl-(trans-4-hydroxycyclohexyl)carbamat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl (trans-4-hydroxycyclohexyl)carbamate, trans-4-Aminocyclohexan-1-ol, N-BOC protected
OR3290-5G
APOSOR3290-5GEA
21.9
EUR
APOSOR3290-5G
2-Methyl-2-propanyl-(trans-4-hydroxycyclohexyl)carbamat 98%
trans-4-(Boc-amino)cyclohexanol
Formel:
C₁₁H₂₁NO₃ Molecular Weight: 215,28 g/mol Schmelzpunkt: 169…173 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03844613 CAS-Nummer: 111300-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...