1-Ethyl-4-(4-nitrophenyl)piperazine 95%

Lieferant: Apollo Scientific

OR32909-1G
APOSOR32909-1GEA 180 EUR
APOSOR32909-1G
1-Ethyl-4-(4-nitrophenyl)piperazine 95%
1-Ethyl-4-(4-nitrophenyl)piperazine
Formel: C₁₂H₁₇N₃O₂
Molecular Weight: 235,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00614372
CAS-Nummer: 115619-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR