Ethyl-1-(4-methoxybenzoyl)-4-piperidinecarboxylate 95%

Lieferant: Apollo Scientific

OR32914-1G OR32914-5G OR32914-25G
APOSOR32914-1GEA 180 EUR
APOSOR32914-1G APOSOR32914-5G APOSOR32914-25G
Ethyl-1-(4-methoxybenzoyl)-4-piperidinecarboxylate 95%
Ethyl-1-(4-methoxybenzoyl)-4-piperidinecarboxylate
Formel: C₁₆H₂₁NO₄
Molecular Weight: 291.35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01213113
CAS-Nummer: 349397-64-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR