5-Amino-3-methyl-1-p-tolylpyrazole 95%

Lieferant: Apollo Scientific

Synonyme: 3-Methyl-1-(4-methylphenyl)-1H-pyrazol-5-amine

OR32925-5G
APOSOR32925-5GEA 97.3 EUR
APOSOR32925-5G
5-Amino-3-methyl-1-p-tolylpyrazole 95%
5-Amino-3-methyl-1-p-tolylpyrazole
Formel: C₁₁H₁₃N₃
Molecular Weight: 187,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00020725
CAS-Nummer: 62535-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR