4-Amino-N-(2-piperidinoethyl)benzenecarboxamide 95%
Lieferant: Apollo Scientific
Synonyme:
4-Amino-N-[2-(piperidin-1-yl)ethyl]benzamide
OR32929-1G
OR32929-5G
APOSOR32929-1GEA
538
EUR
APOSOR32929-1G
APOSOR32929-5G
4-Amino-N-(2-piperidinoethyl)benzenecarboxamide 95%
4-Amino-N-(2-piperidinoethyl)benzenecarboxamide
Formel:
C₁₄H₂₁N₃O Molecular Weight: 247,34 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06808529 CAS-Nummer: 51-08-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...