[4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl]methanol 97%

Lieferant: Apollo Scientific

OR32938-1G OR32938-5G
APOSOR32938-1GEA 180 EUR
APOSOR32938-1G APOSOR32938-5G
[4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl]methanol 97%
[4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl]methanol
Formel: C₁₃H₁₅NO
Molecular Weight: 201.27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04973732
CAS-Nummer: 773870-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR