Methyl-4-n-octyloxybenzoate 99+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-(octyloxy)benzoate

OR3295-100G OR3295-25G OR3295-5G
APOSOR3295-100GEA 77 EUR
APOSOR3295-100G APOSOR3295-25G APOSOR3295-5G
Methyl-4-n-octyloxybenzoate 99+%
Methyl-4-n-octyloxybenzoate
Formel: C₁₆H₂₄O₃
Molecular Weight: 264,360 g/mol
Schmelzpunkt: 35…37 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00070805
CAS-Nummer: 62435-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR