1-(6-Chlor-2-pyridinyl)piperazin 95%

Lieferant: Apollo Scientific

OR32950-1G OR32950-5G
APOSOR32950-1GEA 163 EUR
APOSOR32950-1G APOSOR32950-5G
1-(6-Chlor-2-pyridinyl)piperazin 95%
1-(6-Chlor-2-pyridinyl)piperazin
Formel: C₉H₁₂ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00139009
CAS-Nummer: 87394-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR