5-Isochinolinboronsäure 97%

Lieferant: Apollo Scientific
Achtung

OR3296-100MG OR3296-1G
APOSOR3296-100MGEA 31.9 EUR
APOSOR3296-100MG APOSOR3296-1G
5-Isochinolinboronsäure 97%
5-Isoquinolineboronic acid
Formel: C₉H₈BNO₂
Molecular Weight: 172.98 g/mol
Schmelzpunkt: 186…188 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03839356
CAS-Nummer: 371766-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR