3-(Benzyloxy)-2-methyl-4(1H)-pyridinone 95%
Lieferant: Apollo Scientific
Synonyme:
3-(Benzyloxy)-2-methylpyridin-4(1H)-one
OR32987-1G
OR32987-5G
APOSOR32987-1GEA
101
EUR
APOSOR32987-1G
APOSOR32987-5G
3-(Benzyloxy)-2-methyl-4(1H)-pyridinone 95%
3-(Benzyloxy)-2-methyl-4(1H)-pyridinone
Formel:
C₁₃H₁₃NO₂ Molecular Weight: 215,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00215656 CAS-Nummer: 61160-18-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...