1-Phenylisatin 97%

Lieferant: Apollo Scientific

Synonyme: N-Phenyl-2,3-indolindion, 1-Phenyl-1H-indol-2,3-dion, N-Phenyl-2,3-dioxoindolin, 1-Phenyl-2,3-dioxoindolin, 1-Phenyl-2,3-dihydroindol-2,3-dion, 1-Phenylindolin-2,3-dion, N-Phenyl-1H-indol-2,3-dion, N-Phenylindolin-2,3-dion, N-Phenylisatin, N-Phenyl-2,3-dihydroindol-2,3-dion, 1-Phenyl-2,3-indolindion

OR32991-5G OR32991-25G
APOSOR32991-5GEA 91.5 EUR
APOSOR32991-5G APOSOR32991-25G
1-Phenylisatin 97%
1-Phenylisatin
Formel: C₁₄H₉NO₂
Molecular Weight: 223,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082681
CAS-Nummer: 723-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR