4-Bromo-N-(2-hydroxyethyl)benzenecarboxamide 95%
Lieferant: Apollo Scientific
Synonyme:
4-Bromo-N-(2-hydroxyethyl)benzamide
OR32995-1G
OR32995-25G
OR32995-5G
APOSOR32995-1GEA
180
EUR
APOSOR32995-1G
APOSOR32995-25G
APOSOR32995-5G
4-Bromo-N-(2-hydroxyethyl)benzenecarboxamide 95%
4-Bromo-N-(2-hydroxyethyl)benzenecarboxamide
Formel:
C₉H₁₀BrNO₂ Molecular Weight: 244.09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00456170 CAS-Nummer: 57728-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...