2-[Cyclohexyl(methyl)amino]nicotinic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2-[Cyclohexyl(methyl)amino]pyridine-3-carboxylic acid

OR33054-1G OR33054-5G
APOSOR33054-1GEA 270 EUR
APOSOR33054-1G APOSOR33054-5G
2-[Cyclohexyl(methyl)amino]nicotinic acid 95%
2-[Cyclohexyl(methyl)amino]nicotinic acid
Formel: C₁₃H₁₈N₂O₂
Molecular Weight: 234,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08056648
CAS-Nummer: 886361-91-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR