3-(4-Isopropylphenyl)-2-propen-1-ol 95%
Lieferant: Apollo Scientific
OR33055-1G
OR33055-5G
APOSOR33055-1GEA
336
EUR
APOSOR33055-1G
APOSOR33055-5G
3-(4-Isopropylphenyl)-2-propen-1-ol 95%
3-(4-Isopropylphenyl)-2-propen-1-ol
Formel:
C₁₂H₁₆O Molecular Weight: 176,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08056650 CAS-Nummer: 274907-08-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...