4-Benzyl-1(2H)-phthalazinone 95%

Lieferant: Apollo Scientific

Synonyme: 4-Benzylphthalazin-1(2H)-one

OR33063-1G OR33063-5G OR33063-25G
APOSOR33063-1GEA 180 EUR
APOSOR33063-1G APOSOR33063-5G APOSOR33063-25G
4-Benzyl-1(2H)-phthalazinone 95%
4-Benzyl-1(2H)-phthalazinone
Formel: C₁₅H₁₂N₂O
Molecular Weight: 236.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00139376
CAS-Nummer: 32003-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR