1-(2-Chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: 1-[(2-Chlorophenyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid

OR33070-1G OR33070-5G OR33070-25G
APOSOR33070-1GEA 180 EUR
APOSOR33070-1G APOSOR33070-5G APOSOR33070-25G
1-(2-Chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%
1-(2-Chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid
Formel: C₁₃H₁₀ClNO₃
Molecular Weight: 263,68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01935944
CAS-Nummer: 4399-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR