1-Benzyl-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: 1-Benzyl-6-oxo-1,6-dihydropyridine-3-carboxylic acid

OR33072-1G OR33072-5G
APOSOR33072-1GEA 270 EUR
APOSOR33072-1G APOSOR33072-5G
1-Benzyl-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%
1-Benzyl-6-oxo-1,6-dihydro-3-pyridine carboxylic acid
Formel: C₁₃H₁₁NO₃
Molecular Weight: 229,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00101801
CAS-Nummer: 4332-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR