1-Methyl-3-indolylessigsäure 95%

Lieferant: Apollo Scientific

Synonyme: 1-Methyl-3-indoleacetic acid, 2-(1-Methyl-1H-indol-3-yl)acetic acid

OR33110-25G OR33110-5G
APOSOR33110-25GEA 516 EUR
APOSOR33110-25G APOSOR33110-5G
1-Methyl-3-indolylessigsäure 95%
1-Methyl-3-indolylessigsäure
Formel: C₁₁H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00130160
CAS-Nummer: 1912-48-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR