1-(4-Bromophenyl)-3-(dimethylamino)-2-buten-1-one 97%
Lieferant: Apollo Scientific
OR33118-1G
OR33118-5G
APOSOR33118-1GEA
270
EUR
APOSOR33118-1G
APOSOR33118-5G
1-(4-Bromophenyl)-3-(dimethylamino)-2-buten-1-one 97%
1-(4-Bromophenyl)-3-(dimethylamino)-2-buten-1-one
Formel:
C₁₂H₁₄BrNO Molecular Weight: 268,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08056661 CAS-Nummer: 178261-64-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...