N-(4-Chlorobenzyl)urea 95%

Lieferant: Apollo Scientific

Synonyme: 1-(4-Chlorobenzyl)urea

OR33123-1G OR33123-25G OR33123-5G
APOSOR33123-1GEA 180 EUR
APOSOR33123-1G APOSOR33123-25G APOSOR33123-5G
N-(4-Chlorobenzyl)urea 95%
N-(4-Chlorobenzyl)urea
Formel: C₈H₉ClN₂O
Molecular Weight: 184,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01314630
CAS-Nummer: 65608-74-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR