N-(4-Iodophenyl)cyclopropanecarboxamide 95%
Lieferant: Apollo Scientific
OR33126-1G
OR33126-5G
APOSOR33126-1GEA
180
EUR
APOSOR33126-1G
APOSOR33126-5G
N-(4-Iodophenyl)cyclopropanecarboxamide 95%
N-(4-Iodophenyl)cyclopropanecarboxamide
Formel:
C₁₀H₁₀INO Lagertemperatur: Raumtemperatur |
MDL:
MFCD01327898 CAS-Nummer: 23779-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...