2-(4-Iodophenoxy)pyrimidine 95%

Lieferant: Apollo Scientific

OR33128-1G
APOSOR33128-1GEA 180 EUR
APOSOR33128-1G
2-(4-Iodophenoxy)pyrimidine 95%
2-(4-Iodophenoxy)pyrimidine
Formel: C₁₀H₇IN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD01443687
CAS-Nummer: 330792-86-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR