4-Cyanphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Cyanobenzeneboronic acid, (4-Cyanophenyl)metaboric acid, Dihydroxy(4-cyanophenyl)borane

OR3313-25G OR3313-5G OR3313-100G
APOSOR3313-25GEA 43.5 EUR
APOSOR3313-25G APOSOR3313-5G APOSOR3313-100G
4-Cyanphenylboronsäure
4-Cyanphenylboronsäure
Formel: C₇H₆BNO₂
Molecular Weight: 146,94 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318968
CAS-Nummer: 126747-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR