1-[4-(2-Pyrimidinyloxy)phenyl]-1-ethanone 95%
Lieferant: Apollo Scientific
OR33130-1G
OR33130-25G
OR33130-5G
APOSOR33130-1GEA
180
EUR
APOSOR33130-1G
APOSOR33130-25G
APOSOR33130-5G
1-[4-(2-Pyrimidinyloxy)phenyl]-1-ethanone 95%
1-[4-(2-Pyrimidinyloxy)phenyl]-1-ethanone
Formel:
C₁₂H₁₀N₂O₂ Molecular Weight: 214,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01443772 CAS-Nummer: 182141-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...