3-Amino-3-(4-bromophenyl)acrylonitrile 95%
Lieferant: Apollo Scientific
Synonyme:
(2E)-3-Amino-3-(4-bromophenyl)prop-2-enenitrile
OR33137-1G
OR33137-25G
OR33137-5G
APOSOR33137-1GEA
180
EUR
APOSOR33137-1G
APOSOR33137-25G
APOSOR33137-5G
3-Amino-3-(4-bromophenyl)acrylonitrile 95%
3-Amino-3-(4-bromophenyl)acrylonitrile
Formel:
C₉H₇BrN₂ Molecular Weight: 223,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01871637 CAS-Nummer: 748807-82-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...