N-(3-Acetylphenyl)-3-chloro-2,2-dimethylpropanamide 95%

Lieferant: Apollo Scientific

OR33148-1G OR33148-25G OR33148-5G
APOSOR33148-1GEA 180 EUR
APOSOR33148-1G APOSOR33148-25G APOSOR33148-5G
N-(3-Acetylphenyl)-3-chloro-2,2-dimethylpropanamide 95%
N-(3-Acetylphenyl)-3-chloro-2,2-dimethylpropanamide
Formel: C₁₃H₁₆ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02187344
CAS-Nummer: 453557-74-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR