1-(3-Acetylphenyl)-3,3-dimethyl-2-azetanone 95%

Lieferant: Apollo Scientific

OR33149-1G OR33149-5G
APOSOR33149-1GEA 270 EUR
APOSOR33149-1G APOSOR33149-5G
1-(3-Acetylphenyl)-3,3-dimethyl-2-azetanone 95%
1-(3-Acetylphenyl)-3,3-dimethyl-2-azetanone
Formel: C₁₃H₁₅NO₂
Molecular Weight: 217,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02570851
CAS-Nummer: 453557-75-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR