4-(2-Oxiranylmethoxy)benzenecarbonitrile 95%

Lieferant: Apollo Scientific

Synonyme: 4-[(Oxiran-2-yl)methoxy]benzonitrile

OR33150-1G OR33150-5G
APOSOR33150-1GEA 270 EUR
APOSOR33150-1G APOSOR33150-5G
4-(2-Oxiranylmethoxy)benzenecarbonitrile 95%
4-(2-Oxiranylmethoxy)benzenecarbonitrile
Formel: C₁₀H₉NO₂
Molecular Weight: 175.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04117819
CAS-Nummer: 38791-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR